Assay Details
| Gene Symbol: pigg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026952.4 | NP_001022123.1 | 2 - 3 | 453 | 103 | |
| NM_001381530.2 | NP_001367076.1 | 1 - 2 | 118 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001026952.4, NM_001381530.2 transcripts.
Set Membership: |
|

pigg-1
Genomic Map