Assay Details
| Gene Symbol: pigg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026952.4 | NP_001022123.1 | 4 - 5 | 1493 | 132 | |
| NM_001381530.2 | NP_001367076.1 | 3 - 4 | 1158 | 132 | ||
More Information
Set Membership: |
|

pigg-1
Genomic Map