Assay Details
| Gene Symbol: pigg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026952.4 | NP_001022123.1 | 6 - 7 | 1852 | 83 | |
| NM_001381530.2 | NP_001367076.1 | 5 - 6 | 1517 | 83 | ||
More Information
Set Membership: |
|

pigg-1
Genomic Map