Assay Details
| Gene Symbol: pigg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026952.4 | NP_001022123.1 | 8 - 9 | 2134 | 105 | |
| NM_001381530.2 | NP_001367076.1 | 7 - 8 | 1799 | 105 | ||
More Information
Set Membership: |
|

pigg-1
Genomic Map