Assay Details
| Gene Symbol: rpl-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063422.7 | NP_495823.1 | 3 - 4 | 462 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpl-26| Gene Symbol: rpl-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063422.7 | NP_495823.1 | 3 - 4 | 462 | 75 | |
Set Membership: |
|