Assay Details
| Gene Symbol: cutl-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322749.3 | NP_001309578.1 | 10 | 1781 | 139 | |
| NM_001322750.3 | NP_001309579.1 | 7 - 8 | 1363 | 139 | ||
| NM_001322751.1 | NP_001309580.1 | 1 - 2 | 274 | 139 | ||
More Information
Set Membership: |
|

cutl-12
Genomic Map