Assay Details
| Gene Symbol: pyr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063437.6 | NP_495838.2 | 9 - 10 | 2109 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pyr-1| Gene Symbol: pyr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063437.6 | NP_495838.2 | 9 - 10 | 2109 | 143 | |
Set Membership: |
|