Assay Details
| Gene Symbol: sphk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026846.4 | NP_001022017.1 | 1 - 2 | 241 | 137 | |
| NM_001381535.1 | NP_001367075.1 | 1 - 2 | 90 | 137 | ||
More Information
Set Membership: |
|

sphk-1
Genomic Map