Assay Details
| Gene Symbol: sphk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026846.4 | NP_001022017.1 | 4 - 5 | 827 | 120 | |
| NM_001267250.2 | NP_001254179.1 | 4 - 5 | 866 | 120 | ||
| NM_001267251.3 | NP_001254180.1 | 2 - 3 | 421 | 120 | ||
| NM_001381535.1 | NP_001367075.1 | 4 - 5 | 676 | 120 | ||
More Information
Set Membership: |
|

sphk-1
Genomic Map