Assay Details
| Gene Symbol: cup-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267252.2 | NP_001254181.1 | 2 - 3 | 656 | 95 | |
| NM_001267253.3 | NP_001254182.1 | 1 - 2 | 209 | 95 | ||
More Information
Set Membership: |
|

cup-16
Genomic Map