Assay Details
| Gene Symbol: cyn-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063454.8 | NP_495855.1 | 3 - 4 | 370 | 109 | |
| GenBank mRNA | U34955.1 | - | 2 - 3 | 262 | 109 | |
More Information
Set Membership: |
|

cyn-11
Genomic Map