Assay Details
| Gene Symbol: cyn-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063454.8 | NP_495855.1 | 4 - 5 | 554 | 71 | |
| GenBank mRNA | U34955.1 | - | 3 - 4 | 446 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cyn-11| Gene Symbol: cyn-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063454.8 | NP_495855.1 | 4 - 5 | 554 | 71 | |
| GenBank mRNA | U34955.1 | - | 3 - 4 | 446 | 71 | |
Set Membership: |
|