Assay Details
| Gene Symbol: mak-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027067.5 | NP_001022238.1 | 5 - 6 | 876 | 101 | |
| NM_001373775.3 | NP_001359655.1 | 7 | 1283 | 101 | ||
| NM_001373776.2 | NP_001359681.1 | 7 | 1521 | 101 | ||
| NM_063459.5 | NP_495860.1 | 7 - 8 | 1402 | 101 | ||
More Information
Set Membership: |
|

mak-1
Genomic Map