Assay Details
| Gene Symbol: pah-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267255.3 | NP_001254184.1 | 5 - 6 | 773 | 65 | |
| NM_001267256.2 | NP_001254185.1 | 5 - 6 | 850 | 65 | ||
More Information
Set Membership: |
|

pah-1
Genomic Map