Assay Details
| Gene Symbol: pho-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063474.6 | NP_495875.2 | 6 - 7 | 1238 | 105 | |
| NR_023949.1 | 6 - 7 | 1165 | 105 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pho-13| Gene Symbol: pho-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063474.6 | NP_495875.2 | 6 - 7 | 1238 | 105 | |
| NR_023949.1 | 6 - 7 | 1165 | 105 | |||
Set Membership: |
|