Assay Details
| Gene Symbol: agxt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063484.7 | NP_495885.1 | 8 - 9 | 967 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

agxt-1| Gene Symbol: agxt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063484.7 | NP_495885.1 | 8 - 9 | 967 | 90 | |
Set Membership: |
|