Assay Details
| Gene Symbol: ddx-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267257.3 | NP_001254186.1 | 10 - 11 | 1241 | 92 | |
| NM_001306538.3 | NP_001293467.1 | 5 - 6 | 913 | 92 | ||
| NM_063490.5 | NP_495891.1 | 11 - 12 | 2495 | 92 | ||
| NM_063491.6 | NP_495892.1 | 11 - 12 | 1341 | 92 | ||
| NM_063492.2 | NP_495893.1 | 10 - 11 | 1268 | 92 | ||
More Information
Set Membership: |
|

ddx-19
Genomic Map