Assay Details
| Gene Symbol: ddx-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267257.3 | NP_001254186.1 | 11 - 12 | 1311 | 63 | |
| NM_001306538.3 | NP_001293467.1 | 6 - 7 | 983 | 63 | ||
| NM_063490.5 | NP_495891.1 | 12 - 13 | 2565 | 63 | ||
| NM_063491.6 | NP_495892.1 | 12 - 13 | 1411 | 63 | ||
| NM_063492.2 | NP_495893.1 | 11 - 12 | 1338 | 63 | ||
More Information
Set Membership: |
|

ddx-19
Genomic Map