Assay Details
| Gene Symbol: ddx-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267257.3 | NP_001254186.1 | 6 - 7 | 432 | 61 | |
| NM_001306538.3 | NP_001293467.1 | 1 - 2 | 104 | 61 | ||
| NM_063490.5 | NP_495891.1 | 7 - 8 | 1686 | 61 | ||
| NM_063491.6 | NP_495892.1 | 7 - 8 | 532 | 61 | ||
| NM_063492.2 | NP_495893.1 | 6 - 7 | 459 | 61 | ||
More Information
Set Membership: |
|

ddx-19
Genomic Map