Assay Details
| Gene Symbol: ddx-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267257.3 | NP_001254186.1 | 8 - 9 | 1000 | 89 | |
| NM_001306538.3 | NP_001293467.1 | 3 - 4 | 672 | 89 | ||
| NM_063490.5 | NP_495891.1 | 9 - 10 | 2254 | 89 | ||
| NM_063491.6 | NP_495892.1 | 9 - 10 | 1100 | 89 | ||
| NM_063492.2 | NP_495893.1 | 8 - 9 | 1027 | 89 | ||
More Information
Set Membership: |
|

ddx-19
Genomic Map