Assay Details
| Gene Symbol: tdp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026995.7 | NP_001022166.1 | 2 - 3 | 293 | 99 | |
| NM_001267260.3 | NP_001254189.1 | 2 - 3 | 295 | 99 | ||
| NR_151561.1 | 2 - 3 | 286 | 99 | |||
More Information
Set Membership: |
|

tdp-1
Genomic Map