Assay Details
| Gene Symbol: dep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267263.3 | NP_001254192.1 | 15 - 16 | 3754 | 77 | |
| NM_001267264.3 | NP_001254193.1 | 1 - 2 | 290 | 77 | ||
More Information
Set Membership: |
|

dep-1
Genomic Map