Assay Details
| Gene Symbol: dep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267263.3 | NP_001254192.1 | 16 - 17 | 4206 | 101 | |
| NM_001267264.3 | NP_001254193.1 | 2 - 3 | 742 | 101 | ||
More Information
Set Membership: |
|

dep-1
Genomic Map