Assay Details
| Gene Symbol: znfx-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381556.1 | NP_001367278.1 | 10 - 11 | 3571 | 107 | |
| NM_001381557.1 | NP_001366769.1 | 10 - 11 | 3615 | 107 | ||
More Information
Set Membership: |
|

znfx-1
Genomic Map