Assay Details
| Gene Symbol: aqp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381560.2 | NP_001366692.1 | 2 - 3 | 228 | 65 | |
| NM_001383913.1 | 2 - 3 | 241 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aqp-2| Gene Symbol: aqp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381560.2 | NP_001366692.1 | 2 - 3 | 228 | 65 | |
| NM_001383913.1 | 2 - 3 | 241 | 65 | |||
Set Membership: |
|