Assay Details
| Gene Symbol: wdfy-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063582.4 | NP_495983.1 | 4 - 5 | 746 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wdfy-2| Gene Symbol: wdfy-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063582.4 | NP_495983.1 | 4 - 5 | 746 | 69 | |
Set Membership: |
|