Assay Details
| Gene Symbol: scp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026880.5 | NP_001022051.1 | 10 - 11 | 3064 | 94 | |
| NM_001383916.1 | 10 - 11 | 3056 | 94 | |||
More Information
Set Membership: |
|

scp-1
Genomic Map