Assay Details
| Gene Symbol: scp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026880.5 | NP_001022051.1 | 11 - 12 | 3195 | 88 | |
| NM_001383916.1 | 11 - 12 | 3187 | 88 | |||
More Information
Set Membership: |
|

scp-1
Genomic Map