Assay Details
| Gene Symbol: scp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026880.5 | NP_001022051.1 | 3 - 4 | 690 | 150 | |
| NM_001383916.1 | 3 - 4 | 682 | 150 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383916.1 transcripts.
Set Membership: |
|

scp-1
Genomic Map