Assay Details
| Gene Symbol: scp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026880.5 | NP_001022051.1 | 7 - 8 | 2204 | 74 | |
| NM_001383916.1 | 7 - 8 | 2196 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scp-1| Gene Symbol: scp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026880.5 | NP_001022051.1 | 7 - 8 | 2204 | 74 | |
| NM_001383916.1 | 7 - 8 | 2196 | 74 | |||
Set Membership: |
|