Assay Details
| Gene Symbol: sdhb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063591.7 | NP_495992.1 | 4 - 5 | 589 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sdhb-1| Gene Symbol: sdhb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063591.7 | NP_495992.1 | 4 - 5 | 589 | 64 | |
Set Membership: |
|