Assay Details
| Gene Symbol: zfp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063654.5 | NP_496055.1 | 6 - 7 | 1196 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zfp-2| Gene Symbol: zfp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063654.5 | NP_496055.1 | 6 - 7 | 1196 | 104 | |
Set Membership: |
|