Assay Details
| Gene Symbol: parp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026886.5 | NP_001022057.1 | 1 - 2 | 575 | 142 | |
| GenBank mRNA | AF500111.1 | - | 1 - 2 | 591 | 142 | |
More Information
Set Membership: |
|

parp-2
Genomic Map