Assay Details
| Gene Symbol: parp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026886.5 | NP_001022057.1 | 4 - 5 | 1478 | 86 | |
| GenBank mRNA | AF500111.1 | - | 4 - 5 | 1494 | 86 | |
| Gene Symbol: E02H1.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383918.1 | 3 - 4 | 1565 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383918.1 transcripts.
Set Membership: |
|

parp-2
Genomic Map