Assay Details
| Gene Symbol: arc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063688.8 | NP_496089.3 | 2 - 3 | 770 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

arc-1| Gene Symbol: arc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063688.8 | NP_496089.3 | 2 - 3 | 770 | 83 | |
Set Membership: |
|