Assay Details
| Gene Symbol: chil-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063727.6 | NP_496128.1 | 3 - 4 | 284 | 120 | |
| Gene Symbol: C08H9.18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383925.1 | 3 - 4 | 1043 | 120 | ||
More Information
Set Membership: |
|

chil-3
Genomic Map