Assay Details
| Gene Symbol: old-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063729.4 | NP_496130.2 | 1 - 2 | 105 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_063729.4 transcripts.
Set Membership: |
|

old-1
Genomic Map