Assay Details
| Gene Symbol: ttc-36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063767.4 | NP_496168.1 | 1 - 2 | 220 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ttc-36| Gene Symbol: ttc-36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063767.4 | NP_496168.1 | 1 - 2 | 220 | 75 | |
Set Membership: |
|