Assay Details
| Gene Symbol: mtcu-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063768.7 | NP_496169.1 | 11 - 12 | 2874 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mtcu-2| Gene Symbol: mtcu-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063768.7 | NP_496169.1 | 11 - 12 | 2874 | 70 | |
Set Membership: |
|