Assay Details
| Gene Symbol: flp-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381575.1 | NP_001366711.1 | 2 - 3 | 259 | 102 | |
| GenBank mRNA | AF042390.1 | - | 1 - 2 | 167 | 102 | |
More Information
Set Membership: |
|

flp-4
Genomic Map