Assay Details
| Gene Symbol: puf-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063777.8 | NP_496178.1 | 5 - 6 | 1912 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

puf-12| Gene Symbol: puf-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063777.8 | NP_496178.1 | 5 - 6 | 1912 | 59 | |
Set Membership: |
|