Assay Details
| Gene Symbol: srb-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063798.1 | NP_496199.1 | 6 - 7 | 826 | 95 | |
| Gene Symbol: R05H5.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063799.7 | NP_496200.1 | 1 - 2 | 90 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_063799.7 transcripts.
Set Membership: |
|

srb-6
Genomic Map