Assay Details
| Gene Symbol: atad-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063809.8 | NP_496210.2 | 1 - 2 | 377 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atad-3| Gene Symbol: atad-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063809.8 | NP_496210.2 | 1 - 2 | 377 | 76 | |
Set Membership: |
|