Assay Details
| Gene Symbol: clpp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267310.4 | NP_001254239.1 | 1 - 2 | 193 | 75 | |
| NM_001267311.3 | NP_001254240.1 | 1 - 2 | 142 | 75 | ||
More Information
Set Membership: |
|

clpp-1
Genomic Map