Assay Details
| Gene Symbol: gcy-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063818.1 | NP_496219.1 | 8 - 9 | 1326 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-5| Gene Symbol: gcy-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063818.1 | NP_496219.1 | 8 - 9 | 1326 | 80 | |
Set Membership: |
|