Assay Details
| Gene Symbol: stip-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373788.1 | NP_001359529.1 | 1 - 2 | 123 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

stip-1| Gene Symbol: stip-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373788.1 | NP_001359529.1 | 1 - 2 | 123 | 73 | |
Set Membership: |
|