Assay Details
| Gene Symbol: myrf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063861.5 | NP_496262.2 | 5 - 6 | 2316 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

myrf-1| Gene Symbol: myrf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063861.5 | NP_496262.2 | 5 - 6 | 2316 | 86 | |
Set Membership: |
|