Assay Details
| Gene Symbol: dap-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063879.6 | NP_496280.1 | 2 - 3 | 947 | 88 | |
| GenBank mRNA | AF058329.1 | - | 2 - 3 | 985 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dap-3| Gene Symbol: dap-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063879.6 | NP_496280.1 | 2 - 3 | 947 | 88 | |
| GenBank mRNA | AF058329.1 | - | 2 - 3 | 985 | 88 | |
Set Membership: |
|