Assay Details
| Gene Symbol: algn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063881.6 | NP_496282.2 | 4 - 5 | 1363 | 65 | |
| Gene Symbol: crn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383937.1 | 1 - 2 | 534 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383937.1 transcripts.
Set Membership: |
|

algn-9
Genomic Map