Assay Details
| Gene Symbol: algn-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063881.6 | NP_496282.2 | 5 - 6 | 1665 | 78 | |
| Gene Symbol: crn-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383937.1 | 2 - 3 | 836 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383937.1 transcripts.
Set Membership: |
|

algn-9
Genomic Map